Assay Details
| Gene Symbol: RSPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286506.1 | NP_001273435.1 | 5 - 6 | 630 | 111 | |
| NM_080860.3 | NP_543136.1 | 6 - 7 | 744 | 111 | ||
| XM_005261208.2 | XP_005261265.1 | 4 - 5 | 384 | 111 | ||
| GenBank mRNA | AB006536.1 | - | 6 - 7 | 744 | 111 | |
| AB041016.1 | - | 6 - 7 | 681 | 111 | ||
| AK057315.1 | - | 5 - 6 | 2195 | 111 | ||
| AK314746.1 | - | 6 - 7 | 581 | 111 | ||
| BC101519.1 | - | 6 - 7 | 623 | 8069025 | 111 | |
| BC113367.1 | - | 6 - 7 | 623 | 8322419 | 111 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000489.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR21005929.hg.1 | ENST00000291536.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21005929.hg.1 | ENST00000398352.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21005929.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21005929.hg.1 | TC21000489.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000398352.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000398352.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000398352.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000398352.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000398352.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000398352.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000398352.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000398352.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000398352.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000398352.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100155969.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100155969.hg.1 | ENST00000398352.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100155969.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100155969.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100155969.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100155969.hg.1 | TC2100008256.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100155969.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100155969.hg.1 | ENST00000398352.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100155969.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100155969.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100155969.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100155968.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100155968.hg.1 | ENST00000398352.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100155968.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100155968.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100155968.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100155968.hg.1 | TC2100008256.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100155968.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100155968.hg.1 | ENST00000398352.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100155968.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100155968.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100155968.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000291536.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000398352.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000398352.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000398352.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000398352.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000398352.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000398352.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000398352.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000398352.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000398352.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000398352.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | ENST00000493019.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_001286506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_080860.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008256.hg.1 | NM_080860.3 | Refseq |

RSPH1
Genomic Map