Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000520568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000520568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000520568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000520568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000520568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000522726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000522726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000522726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000522726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000522726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000544227.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000544227.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000544227.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000544227.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000544227.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000544227.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000544227.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000544227.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
ENST00000544227.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018069.hg.1 |
TC08001396.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018069.hg.1 |
ENST00000286614.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08018069.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000520568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000520568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000520568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000520568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000520568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000522726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000522726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000522726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000522726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000522726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000544227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000544227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000544227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000544227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000544227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000544227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000544227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000544227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
ENST00000544227.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011041.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175040.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175040.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175040.hg.1 |
ENST00000286614.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175040.hg.1 |
NM_005941.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175040.hg.1 |
TC0800011041.hg.1 |
Clariom D TC |