Assay Details
| Gene Symbol: PPP1R12C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271618.1 | NP_001258547.1 | 10 - 11 | 1441 | 70 | |
| NM_017607.3 | NP_060077.1 | 10 - 11 | 1444 | 70 | ||
| XM_005259013.3 | XP_005259070.1 | 10 - 11 | 1435 | 70 | ||
| XM_011527045.2 | XP_011525347.1 | 10 - 11 | 1433 | 70 | ||
| GenBank mRNA | AB209452.1 | - | 10 - 11 | 1254 | 70 | |
| AF312028.1 | - | 10 - 11 | 1405 | 70 | ||
| AK027086.1 | - | 10 - 11 | 1079 | 70 | ||
| AK297187.1 | - | 7 - 8 | 727 | 70 | ||
| AK297426.1 | - | 10 - 11 | 1293 | 70 | ||
| AL137618.1 | - | 5 - 6 | 478 | 70 | ||
| BC008030.1 | - | 1 - 2 | 131 | 70 | ||
| BC010628.1 | - | 9 - 10 | 1222 | 70 | ||
More Information
Set Membership: |
|

PPP1R12C
Genomic Map