Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003009.hg.1 |
ENST00000361866.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003009.hg.1 |
ENST00000538397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003009.hg.1 |
ENST00000546256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003009.hg.1 |
NM_001848.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003009.hg.1 |
TC21000249.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2100150437.hg.1 |
ENST00000361866.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150437.hg.1 |
ENST00000612273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150437.hg.1 |
NM_001848.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100150437.hg.1 |
TC2100007446.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100150437.hg.1 |
ENST00000361866.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150437.hg.1 |
ENST00000612273.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150437.hg.1 |
NM_001848.2 |
Refseq |