Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002262.hg.1 |
ENST00000340648.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002262.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002262.hg.1 |
TC21000212.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
ENST00000470886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
ENST00000470886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
ENST00000470886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
ENST00000470886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
ENST00000470886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
ENST00000470886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
ENST00000470886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
ENST00000470886.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100149323.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100149323.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100149323.hg.1 |
TC2100007318.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100149323.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100149323.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
ENST00000340648.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
ENST00000470886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
ENST00000470886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
ENST00000470886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
ENST00000470886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
ENST00000470886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
ENST00000470886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
ENST00000470886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
ENST00000470886.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007318.hg.1 |
NM_015056.2 |
Refseq |