| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000382760.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000382760.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000382760.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000382760.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000382760.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000382760.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000382760.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000486043.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000486043.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000486043.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000486043.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000498140.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000498140.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000498140.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000498140.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000498140.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000498140.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000498140.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000498140.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
ENST00000498140.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22002812.hg.1 |
ENST00000326010.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22002812.hg.1 |
ENST00000413389.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22002812.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22002812.hg.1 |
TC22000149.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22002813.hg.1 |
ENST00000326010.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22002813.hg.1 |
ENST00000413389.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22002813.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22002813.hg.1 |
TC22000149.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000382760.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000382760.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000382760.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000382760.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000382760.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000382760.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000382760.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000486043.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000486043.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000486043.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000486043.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000498140.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000498140.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000498140.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000498140.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000498140.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000498140.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000498140.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000498140.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
ENST00000498140.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200006894.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200147752.hg.1 |
TC2200006894.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200147752.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200147752.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200147752.hg.1 |
NM_016327.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200147752.hg.1 |
ENST00000326010.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200147752.hg.1 |
ENST00000415388.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200147752.hg.1 |
NM_016327.2 |
Refseq |