Assay Details
| Gene Symbol: MOB3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK074231.1 | - | 1 | 586 | 100 | |
| AK172755.1 | - | 1 | 586 | 100 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

MOB3B
Genomic Map