Assay Details
| Gene Symbol: MOB3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK022201.1 | - | 1 | 673 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | equatorin | EQTN |
AFAF, C9orf11, SPACA8 |
-37.7kb | Sense | Chr. 9 - 27284654 - 27297139 on NCBI Build GRCh38 |
| Downstream | interferon kappa | IFNK |
IFNT1, INFE1 |
186.5kb | Anti-sense | Chr. 9 - 27524314 - 27526498 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

MOB3B
Genomic Map