Assay Details
| Gene Symbol: PITPNM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020845.2 | NP_065896.1 | 15 - 16 | 2542 | 108 | |
| GenBank mRNA | AF334585.1 | - | 14 - 15 | 2481 | 108 | |
| AL133612.1 | - | 6 - 7 | 989 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

PITPNM2
Genomic Map