Assay Details
| Gene Symbol: CDH22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021248.2 | NP_067071.1 | 2 - 3 | 905 | 80 | |
| XM_011528994.2 | XP_011527296.1 | 2 - 3 | 864 | 80 | ||
| GenBank mRNA | AK289584.1 | - | 2 - 3 | 905 | 80 | |
| BC136526.1 | - | 1 - 2 | 654 | 9020516 | 80 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000898.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20012274.hg.1 | ENST00000372262.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012274.hg.1 | ENST00000474438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012274.hg.1 | ENST00000537909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012274.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012274.hg.1 | TC20000898.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012279.hg.1 | ENST00000372262.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012279.hg.1 | ENST00000537909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012279.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012279.hg.1 | TC20000898.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000474438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000474438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000474438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000474438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000474438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000474438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012278.hg.1 | ENST00000372262.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012278.hg.1 | ENST00000537909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012278.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012278.hg.1 | TC20000898.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012277.hg.1 | ENST00000372262.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012277.hg.1 | ENST00000537909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012277.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012277.hg.1 | TC20000898.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2000164858.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000164858.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000164858.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000164858.hg.1 | TC2000009263.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000164858.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000164858.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000164858.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000164856.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000164856.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000164856.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000164856.hg.1 | TC2000009263.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000164856.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000164856.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000164856.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000474438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000474438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000474438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000474438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000474438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000474438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009263.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000164857.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000164857.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000164857.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000164857.hg.1 | TC2000009263.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000164857.hg.1 | ENST00000372262.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000164857.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000164857.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000164861.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000164861.hg.1 | NM_021248.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000164861.hg.1 | TC2000009263.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000164861.hg.1 | ENST00000537909.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000164861.hg.1 | NM_021248.2 | Refseq |

CDH22
Genomic Map