Assay Details
| Gene Symbol: TGM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001323316.1 | NP_001310245.1 | 13 - 14 | 2120 | 130 | |
| NM_001323317.1 | NP_001310246.1 | 11 - 12 | 1894 | 130 | ||
| NM_001323318.1 | NP_001310247.1 | 11 - 12 | 1957 | 130 | ||
| NM_004613.3 | NP_004604.2 | 12 - 13 | 2137 | 130 | ||
| XM_011529028.1 | XP_011527330.1 | 13 - 14 | 2082 | 130 | ||
| GenBank mRNA | AK126508.1 | - | 2 - 3 | 1551 | 130 | |
| AK291714.1 | - | 12 - 13 | 1978 | 130 | ||
| AK295775.1 | - | 11 - 12 | 1735 | 130 | ||
| AK300292.1 | - | 11 - 12 | 1798 | 130 | ||
| AK314618.1 | - | 12 - 13 | 1978 | 130 | ||
| AY675221.1 | - | 11 - 12 | 1906 | 130 | ||
| BC073946.1 | - | 1 - 2 | 125 | 130 | ||
| BQ430537.1 | - | 1 - 2 | 121 | 130 | ||
| BQ938957.1 | - | 4 - 5 | 698 | 130 | ||
| EU794665.1 | - | 12 - 13 | 2006 | 130 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000833.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
phospholipase C-activating G-protein coupled receptor signaling pathway
peptide cross-linking
negative regulation of endoplasmic reticulum calcium ion concentration
positive regulation of apoptotic process
apoptotic cell clearance
positive regulation of cell adhesion
positive regulation of mitochondrial calcium ion concentration
branching involved in salivary gland morphogenesis
salivary gland cavitation
protein-glutamine gamma-glutamyltransferase activity
protein binding
metal ion binding
phospholipase C-activating G-protein coupled receptor signaling pathway
peptide cross-linking
negative regulation of endoplasmic reticulum calcium ion concentration
positive regulation of apoptotic process
apoptotic cell clearance
positive regulation of cell adhesion
positive regulation of mitochondrial calcium ion concentration
branching involved in salivary gland morphogenesis
salivary gland cavitation
protein-glutamine gamma-glutamyltransferase activity
protein binding
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20011450.hg.1 | ENST00000361475.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011450.hg.1 | ENST00000485572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011450.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011450.hg.1 | TC20000833.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011452.hg.1 | ENST00000361475.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011452.hg.1 | ENST00000536701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011452.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011452.hg.1 | TC20000833.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011451.hg.1 | ENST00000361475.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011451.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011451.hg.1 | TC20000833.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000373403.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000373403.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000373403.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000373403.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000373403.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000373403.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000373403.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000373403.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000373403.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000453095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000453095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000453095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000453095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000453095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000453095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000468262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000468262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000468262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000468262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000468262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000468262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000468262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000468262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000468262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000468262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000468262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000468262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000468262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000469269.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000469269.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000469269.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000469269.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000469269.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000474777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000474777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000474777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000474777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000474777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000474777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000474777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | ENST00000474777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_198951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_198951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_198951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_198951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_198951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_198951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_198951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_198951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_198951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_198951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_198951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_198951.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009058.hg.1 | NM_198951.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000163481.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000163481.hg.1 | ENST00000469269.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000163481.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000163481.hg.1 | TC2000009058.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000163481.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000163481.hg.1 | ENST00000469269.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000163481.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000361475.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000373403.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000373403.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000373403.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000373403.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000373403.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000373403.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000373403.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000373403.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000373403.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000453095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000453095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000453095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000453095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000453095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000453095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000468262.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000468262.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000468262.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000468262.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000468262.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000468262.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000468262.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000468262.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000468262.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000468262.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000468262.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000468262.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000468262.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000469269.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000469269.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000469269.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000469269.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000469269.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000474777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000474777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000474777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000474777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000474777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000474777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000474777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | ENST00000474777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_004613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_198951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_198951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_198951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_198951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_198951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_198951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_198951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_198951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_198951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_198951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_198951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_198951.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009058.hg.1 | NM_198951.2 | Refseq |

TGM2
Genomic Map