Assay Details
Gene Symbol: ERICH6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182542.2 | NP_872348.2 | 7 - 8 | 115 | ||
XM_011534965.2 | XP_011533267.1 | 6 - 7 | 115 | |||
XM_017020418.1 | XP_016875907.1 | 6 - 7 | 115 | |||
GenBank mRNA | AK057244.1 | - | 7 - 8 | 115 | ||
BC131494.1 | - | 6 - 7 | 1060 | 115 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC13000635.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR13007205.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13007205.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13007205.hg.1 | TC13000635.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000378977.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000378977.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000378977.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000378977.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000378977.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000378977.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000378977.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000378977.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000378977.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000378977.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000378977.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000482992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000482992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000482992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000482992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000482992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000482992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000504261.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000504261.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000504261.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | ENST00000504261.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000378977.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000378977.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000378977.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000378977.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000378977.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000378977.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000378977.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000378977.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000378977.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000378977.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000378977.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000482992.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000482992.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000482992.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000482992.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000482992.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000482992.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000504261.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000504261.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000504261.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | ENST00000504261.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008827.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300159547.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300159547.hg.1 | ENST00000378977.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300159547.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300159547.hg.1 | ENST00000298738.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300159547.hg.1 | ENST00000378977.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300159547.hg.1 | NM_182542.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300159547.hg.1 | TC1300008827.hg.1 | Clariom D TC |
