Assay Details
Gene Symbol: AGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033655.1 | 5 - 6 | 753 | 114 | ||
GenBank mRNA | CX869892.1 | - | 5 - 6 | 630 | 114 | |
M64076.1 | - | 3 - 4 | 321 | 114 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001769.hg.1
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000502310.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000502310.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000502310.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000502310.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000502310.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000502310.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000502310.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000506853.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000506853.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000506853.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000506853.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000506853.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000506853.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000506853.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000506853.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000506853.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000506853.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000510635.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000510635.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000510635.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000510635.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000510635.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000510635.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000510635.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000510955.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000510955.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000510955.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000510955.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000510955.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000510955.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000510955.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000511231.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000511231.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000511231.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | ENST00000511231.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04024355.hg.1 | TC04001769.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04024355.hg.1 | ENST00000264595.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04024355.hg.1 | ENST00000506853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04024355.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04024355.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04024355.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400185678.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400185678.hg.1 | ENST00000502310.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400185678.hg.1 | ENST00000506853.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400185678.hg.1 | ENST00000510635.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400185678.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400185678.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400185678.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400185678.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400185678.hg.1 | ENST00000502310.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400185678.hg.1 | ENST00000506853.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400185678.hg.1 | ENST00000510635.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400185678.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400185678.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400185678.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400185678.hg.1 | TC0400012496.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000264595.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000502310.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000502310.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000502310.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000502310.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000502310.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000502310.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000502310.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000506853.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000506853.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000506853.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000506853.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000506853.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000506853.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000506853.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000506853.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000506853.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000506853.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000510635.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000510635.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000510635.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000510635.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000510635.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000510635.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000510635.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000510955.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000510955.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000510955.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000510955.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000510955.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000510955.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000510955.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000511231.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000511231.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000511231.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | ENST00000511231.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_000027.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NM_001171988.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NR_033655.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012496.hg.1 | NR_033655.1 | Refseq |
