Assay Details
Gene Symbol: IFI27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001288957.1 | NP_001275886.1 | 1 - 2 | 169 | 91 | |
GenBank mRNA | BC015492.1 | - | 1 - 2 | 169 | 3915505 | 91 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC015492.1,NM_001288957.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
activation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of protein export from nucleus
type I interferon signaling pathway
apoptotic signaling pathway
RNA polymerase II activating transcription factor binding
molecular_function
lamin binding
negative regulation of transcription from RNA polymerase II promoter
activation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of protein export from nucleus
type I interferon signaling pathway
apoptotic signaling pathway
RNA polymerase II activating transcription factor binding
molecular_function
lamin binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000555081.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000555081.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000555081.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000555081.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000555819.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000555819.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000555819.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000555819.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000555819.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000557700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000557700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000557700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000557700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000611954.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000611954.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000611954.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612813.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612813.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612813.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612813.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612813.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612813.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612813.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000612813.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614288.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614288.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614288.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614288.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614288.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614288.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000614648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000616764.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000616764.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000616764.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000616764.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000616764.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000616764.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000616764.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000616764.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000616764.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000618200.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000618200.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000618200.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000618200.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000618200.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000618863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000618863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000618863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000618863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000618863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000618863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620396.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620396.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620396.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620396.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620396.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620396.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000620396.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000621160.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000621160.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000621160.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000621160.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000621160.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000621160.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000621160.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | ENST00000621160.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288952.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288954.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288954.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288954.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288954.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288954.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288954.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288954.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288954.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288954.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288956.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288956.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288956.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288956.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288956.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288956.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288956.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288957.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288957.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288957.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288957.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288957.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288957.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288957.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288957.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288958.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288958.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288958.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288958.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288958.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288958.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288959.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288960.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_001288995.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_005532.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_005532.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_005532.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_005532.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_005532.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_005532.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_005532.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008056.hg.1 | NM_005532.4 | Refseq |
