Assay Details
| Gene Symbol: SERPINA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006220.2 | NP_006211.2 | 925 | 128 | ||
| GenBank mRNA | JX680599.1 | - | 921 | 128 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SERPINA2| Gene Symbol: SERPINA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006220.2 | NP_006211.2 | 925 | 128 | ||
| GenBank mRNA | JX680599.1 | - | 921 | 128 | ||
Set Membership: |
|