Assay Details
| Gene Symbol: TMEM30A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143958.1 | NP_001137430.1 | 1 - 2 | 510 | 117 | |
| GenBank mRNA | AK292823.1 | - | 1 - 2 | 375 | 117 | |
| BC009006.1 | - | 1 - 2 | 361 | 4154200 | 117 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001881.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06022931.hg.1 | TC06001881.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022931.hg.1 | ENST00000230461.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022931.hg.1 | ENST00000370050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022931.hg.1 | ENST00000475111.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022931.hg.1 | ENST00000545449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022931.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022931.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000518161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000518161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000518161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000518161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | ENST00000518161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000518161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000518161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000518161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000518161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | ENST00000518161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012316.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600184977.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600184977.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600184977.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600184977.hg.1 | ENST00000518161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600184977.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600184977.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600184977.hg.1 | ENST00000230461.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600184977.hg.1 | ENST00000370050.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600184977.hg.1 | ENST00000475111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600184977.hg.1 | ENST00000518161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600184977.hg.1 | NM_001143958.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600184977.hg.1 | NM_018247.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600184977.hg.1 | TC0600012316.hg.1 | Clariom D TC |

TMEM30A
Genomic Map