Assay Details
Gene Symbol: EBNA1BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159936.1 | NP_001153408.1 | 6 - 7 | 856 | 67 | |
NM_006824.2 | NP_006815.2 | 5 - 6 | 739 | 67 | ||
GenBank mRNA | AB451256.1 | - | 5 - 6 | 542 | 67 | |
AB451381.1 | - | 5 - 6 | 542 | 67 | ||
AK223455.1 | - | 5 - 6 | 722 | 67 | ||
AK307440.1 | - | 5 - 6 | 685 | 67 | ||
BC009175.2 | - | 5 - 6 | 680 | 2966681 | 67 | |
BC073924.1 | - | 11 - 12 | 1091 | 67 | ||
BT006920.1 | - | 5 - 6 | 542 | 67 | ||
CR456975.1 | - | 5 - 6 | 542 | 67 | ||
U86602.1 | - | 5 - 6 | 684 | 67 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01002580.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000463906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000463906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000463906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000463906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000463906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000463906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000463906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000463906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000463906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000463906.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000472982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000472982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000472982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000472982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000472982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000472982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000472982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000472982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000483082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000483082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000483082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000483082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000483082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000483082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000483082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000483082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000483082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000491223.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000491223.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000491223.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000491223.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000491223.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000491223.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000491223.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | ENST00000491223.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018435.hg.1 | NR_106791.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01040100.hg.1 | TC01002580.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01040100.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01040100.hg.1 | ENST00000431635.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01040100.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01040100.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000463906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000463906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000463906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000463906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000463906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000463906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000463906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000463906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000463906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000463906.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000472982.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000472982.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000472982.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000472982.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000472982.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000472982.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000472982.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000472982.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000483082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000483082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000483082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000483082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000483082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000483082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000483082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000483082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000483082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000491223.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000491223.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000491223.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000491223.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000491223.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000491223.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000491223.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | ENST00000491223.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018435.hg.1 | NR_106791.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100241877.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100241877.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100241877.hg.1 | ENST00000463906.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100241877.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100241877.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100241877.hg.1 | ENST00000236051.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100241877.hg.1 | ENST00000431635.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100241877.hg.1 | ENST00000463906.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100241877.hg.1 | NM_001159936.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100241877.hg.1 | NM_006824.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100241877.hg.1 | TC0100018435.hg.1 | Clariom D TC |
