Assay Details
| Gene Symbol: NPAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321307.1 | NP_001308236.1 | 8 - 9 | 820 | 124 | |
| NM_002519.2 | NP_002510.2 | 8 - 9 | 820 | 124 | ||
| XM_011542854.2 | XP_011541156.1 | 8 - 9 | 820 | 124 | ||
| XM_017017802.1 | XP_016873291.1 | 8 - 9 | 1558 | 124 | ||
| XM_017017803.1 | XP_016873292.1 | 8 - 9 | 1001 | 124 | ||
| GenBank mRNA | AK290020.1 | - | 8 - 9 | 820 | 124 | |
| AK291687.1 | - | 8 - 9 | 744 | 124 | ||
| BC040356.1 | - | 8 - 9 | 811 | 124 | ||
| BC050561.2 | - | 8 - 9 | 805 | 124 | ||
| BC136270.1 | - | 8 - 9 | 741 | 9020257 | 124 | |
| D83243.1 | - | 8 - 9 | 784 | 124 | ||
| U58852.1 | - | 8 - 9 | 718 | 124 | ||
| X97186.1 | - | 8 - 9 | 752 | 124 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11002264.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription involved in G1/S transition of mitotic cell cycle
transcription, DNA-templated
regulation of gene expression
positive regulation of transcription, DNA-templated
negative regulation of nucleic acid-templated transcription
transcription coactivator activity
transcription corepressor activity
protein binding
protein C-terminus binding
protein N-terminus binding
regulation of transcription involved in G1/S transition of mitotic cell cycle
transcription, DNA-templated
regulation of gene expression
positive regulation of transcription, DNA-templated
negative regulation of nucleic acid-templated transcription
transcription coactivator activity
transcription corepressor activity
protein binding
protein C-terminus binding
protein N-terminus binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000527296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000527296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000527296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000527296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000530859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000530859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000530859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000530859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000530859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000530859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000530859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000530859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000530859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000530859.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000530926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000530926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000530926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000530926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000531384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000531384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000531384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000531384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000531384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000531384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000531384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11027137.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11027137.hg.1 | ENST00000533535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11027137.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11027137.hg.1 | TC11002264.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100194025.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100194025.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100194025.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100194025.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100194025.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100194025.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100194025.hg.1 | TC1100012225.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000278612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000527296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000527296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000527296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000527296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000530859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000530859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000530859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000530859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000530859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000530859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000530859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000530859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000530859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000530859.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000530926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000530926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000530926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000530926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000531384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000531384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000531384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000531384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000531384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000531384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000531384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | ENST00000610253.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | NM_002519.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012225.hg.1 | NM_002519.2 | Refseq |

NPAT
Genomic Map