Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025942.hg.1 |
ENST00000377560.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025942.hg.1 |
ENST00000430131.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025942.hg.1 |
ENST00000550722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025942.hg.1 |
NM_001109662.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025942.hg.1 |
TC12001978.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200193115.hg.1 |
ENST00000377560.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193115.hg.1 |
ENST00000550722.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193115.hg.1 |
NM_001109662.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193115.hg.1 |
ENST00000377560.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193115.hg.1 |
ENST00000550722.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193115.hg.1 |
NM_001109662.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193115.hg.1 |
TC1200011968.hg.1 |
Clariom D TC |