Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024403.hg.1 |
TC06001970.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024403.hg.1 |
ENST00000369162.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024403.hg.1 |
NM_006828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187415.hg.1 |
ENST00000369162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187415.hg.1 |
NM_006828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187415.hg.1 |
ENST00000369162.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187415.hg.1 |
NM_006828.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187415.hg.1 |
TC0600012671.hg.1 |
Clariom D TC |