Assay Details
Gene Symbol: MPHOSPH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005792.2 | NP_005783.2 | 2 - 3 | 208 | 152 | |
XM_011522808.2 | XP_011521110.1 | 3 - 4 | 488 | 152 | ||
XM_017022852.1 | XP_016878341.1 | 2 - 3 | 322 | 152 | ||
GenBank mRNA | AK222667.1 | - | 2 - 3 | 208 | 152 | |
AK314163.1 | - | 2 - 3 | 208 | 152 | ||
AW175781.1 | - | 2 - 3 | 240 | 152 | ||
BC005242.1 | - | 2 - 3 | 179 | 3997566 | 152 | |
BC011020.1 | - | 2 - 3 | 185 | 4287267 | 152 | |
BC031017.1 | - | 2 - 3 | 180 | 4670801 | 152 | |
BU595100.1 | - | 2 - 3 | 204 | 152 | ||
X98263.1 | - | 2 - 3 | 191 | 152 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16001302.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR16018080.hg.1 | ENST00000258169.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16018080.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16018080.hg.1 | TC16001302.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000567729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000567729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000567729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000567729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000567729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000568016.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000568016.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000568016.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000568016.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000568016.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000569021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000569021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000569021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000569021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000567729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000567729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000567729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000567729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000567729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000568016.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000568016.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000568016.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000568016.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000568016.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000569021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000569021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000569021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000569021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011007.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600185565.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185565.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185565.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185565.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185565.hg.1 | ENST00000568016.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185565.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185565.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600185565.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185565.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185565.hg.1 | ENST00000563504.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185565.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185565.hg.1 | ENST00000568016.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185565.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185565.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600185565.hg.1 | TC1600011007.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600185569.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185569.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185569.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185569.hg.1 | ENST00000567729.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185569.hg.1 | ENST00000568016.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185569.hg.1 | ENST00000569021.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185569.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185569.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600185569.hg.1 | ENST00000258169.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185569.hg.1 | ENST00000563100.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185569.hg.1 | ENST00000566748.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185569.hg.1 | ENST00000567729.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185569.hg.1 | ENST00000568016.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185569.hg.1 | ENST00000569021.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185569.hg.1 | ENST00000569599.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600185569.hg.1 | NM_005792.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600185569.hg.1 | TC1600011007.hg.1 | Clariom D TC |
