Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018079.hg.1 |
ENST00000258169.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018079.hg.1 |
NM_005792.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018079.hg.1 |
TC16001302.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000258169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000258169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000258169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000258169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000258169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000258169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000258169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000258169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000258169.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000563100.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000563100.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000563100.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000563100.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000563100.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000563100.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000563100.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000563100.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000563100.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000563504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000563504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000563504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000563504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000563504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000563504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000563504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000563504.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000566748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000566748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000566748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000566748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000566748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000566748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000566748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000566748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000566748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000567729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000567729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000567729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000567729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000567729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000568016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000568016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000568016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000568016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000568016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000569021.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000569021.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000569021.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000569021.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000569599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000569599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000569599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000569599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000569599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000569599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000569599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
ENST00000569599.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
NM_005792.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
NM_005792.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
NM_005792.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
NM_005792.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
NM_005792.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
NM_005792.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
NM_005792.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
NM_005792.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011007.hg.1 |
NM_005792.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000258169.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000258169.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000258169.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000258169.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000258169.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000258169.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000258169.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000258169.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000258169.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000563100.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000563100.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000563100.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000563100.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000563100.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000563100.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000563100.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000563100.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000563100.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000563504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000563504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000563504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000563504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000563504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000563504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000563504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000563504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000566748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000566748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000566748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000566748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000566748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000566748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000566748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000566748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000566748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000567729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000567729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000567729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000567729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000567729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000568016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000568016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000568016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000568016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000568016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000569021.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000569021.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000569021.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000569021.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000569599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000569599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000569599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000569599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000569599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000569599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000569599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
ENST00000569599.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
NM_005792.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
NM_005792.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
NM_005792.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
NM_005792.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
NM_005792.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
NM_005792.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
NM_005792.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
NM_005792.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011007.hg.1 |
NM_005792.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600185558.hg.1 |
ENST00000258169.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600185558.hg.1 |
ENST00000563100.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600185558.hg.1 |
ENST00000563504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600185558.hg.1 |
NM_005792.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600185558.hg.1 |
ENST00000258169.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600185558.hg.1 |
ENST00000563100.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600185558.hg.1 |
ENST00000563504.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600185558.hg.1 |
NM_005792.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600185558.hg.1 |
TC1600011007.hg.1 |
Clariom D TC |