Assay Details
Gene Symbol: DHX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001358.2 | NP_001349.2 | 9 - 10 | 1762 | 135 | |
GenBank mRNA | AB001636.1 | - | 9 - 10 | 1755 | 135 | |
AF279891.1 | - | 9 - 10 | 1594 | 135 | ||
AK303506.1 | - | 9 - 10 | 1702 | 135 | ||
BC035974.1 | - | 9 - 10 | 1749 | 4806964 | 135 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001086.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04014934.hg.1 | TC04001086.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04014934.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04014934.hg.1 | ENST00000535946.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04014934.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000476316.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000503562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000503562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000503562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000503562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000504279.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000504279.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000504279.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000504279.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000508368.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000508368.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000508368.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000508368.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000510645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000510645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000510645.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000511553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000511553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000511553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000511553.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000513092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000513092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000513092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | ENST00000513092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400170152.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400170152.hg.1 | ENST00000503562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400170152.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400170152.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400170152.hg.1 | ENST00000503562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400170152.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400170152.hg.1 | TC0400010251.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000336812.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000476316.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000503562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000503562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000503562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000503562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000504279.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000504279.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000504279.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000504279.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000508032.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000508368.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000508368.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000508368.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000508368.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000510645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000510645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000510645.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000511553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000511553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000511553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000511553.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000512903.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000513036.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000513092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000513092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000513092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | ENST00000513092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010251.hg.1 | NM_001358.2 | Refseq |
