Assay Details
| Gene Symbol: EMC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK222615.1 | - | 4 - 5 | 326 | 101 | |
| AK290974.1 | - | 4 - 5 | 312 | 101 | ||
| BC020753.1 | - | 4 - 5 | 324 | 4809693 | 101 | |
| BC021667.1 | - | 4 - 5 | 341 | 3682913 | 101 | |
| CR457402.1 | - | 4 - 5 | 302 | 101 | ||
| D14659.1 | - | 4 - 5 | 308 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000517593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000517593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000517593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000517593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000517593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000517593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000517593.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000517784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000517784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000517784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000517784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000517784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000517784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000517784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000517784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000517784.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000519450.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000519450.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000519450.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000519450.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000519450.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000519450.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000519450.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000519450.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000519642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000519642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000519642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000519642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000519642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000519642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000519642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000520294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000520294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000520294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000520294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000520294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000520294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000520294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000520294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000520294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000524143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000524143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000524143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000524143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000524143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | ENST00000524143.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000517593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000517593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000517593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000517593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000517593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000517593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000517593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000517784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000517784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000517784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000517784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000517784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000517784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000517784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000517784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000517784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000519450.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000519450.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000519450.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000519450.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000519450.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000519450.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000519450.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000519450.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000519642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000519642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000519642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000519642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000519642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000519642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000519642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000520294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000520294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000520294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000520294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000520294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000520294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000520294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000520294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000520294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000524143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000524143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000524143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000524143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000524143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | ENST00000524143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800008531.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800158570.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800158570.hg.1 | ENST00000517593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800158570.hg.1 | ENST00000517784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800158570.hg.1 | ENST00000519642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800158570.hg.1 | ENST00000524143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800158570.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800158570.hg.1 | ENST00000220853.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800158570.hg.1 | ENST00000517593.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800158570.hg.1 | ENST00000517784.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800158570.hg.1 | ENST00000519642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800158570.hg.1 | ENST00000524143.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800158570.hg.1 | NM_014673.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800158570.hg.1 | TC0800008531.hg.1 | Clariom D TC |

EMC2
Genomic Map