Assay Details
Gene Symbol: UBASH3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032873.4 | NP_116262.2 | 7 - 8 | 1473 | 99 | |
XM_005271712.3 | XP_005271769.1 | 7 - 8 | 3719 | 99 | ||
XM_011543041.2 | XP_011541343.1 | 7 - 8 | 1111 | 99 | ||
XM_011543042.1 | XP_011541344.1 | 7 - 8 | 1477 | 99 | ||
GenBank mRNA | AB075839.1 | - | 7 - 8 | 1325 | 99 | |
AF425252.1 | - | 5 - 6 | 770 | 99 | ||
AK075203.1 | - | 7 - 8 | 1441 | 99 | ||
AK222843.1 | - | 7 - 8 | 1423 | 99 | ||
AK222846.1 | - | 7 - 8 | 1423 | 99 | ||
BC007541.1 | - | 7 - 8 | 1427 | 2958242 | 99 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11001115.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of protein kinase activity
negative regulation of signal transduction
peptidyl-tyrosine dephosphorylation
negative regulation of osteoclast differentiation
negative regulation of bone resorption
regulation of release of sequestered calcium ion into cytosol
negative regulation of platelet aggregation
protein tyrosine phosphatase activity
protein binding
negative regulation of protein kinase activity
negative regulation of signal transduction
peptidyl-tyrosine dephosphorylation
negative regulation of osteoclast differentiation
negative regulation of bone resorption
regulation of release of sequestered calcium ion into cytosol
negative regulation of platelet aggregation
protein tyrosine phosphatase activity
protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000525711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000525711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000525711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000526386.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000526386.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000526386.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000526386.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000526493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000526493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000526493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000526493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000529998.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000529998.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000529998.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000529998.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000529998.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000530578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000530578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000530578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000530578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000530917.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000530917.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000530917.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000530917.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000530917.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000530917.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000530917.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000533451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000533451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000533451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000533451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | ENST00000533451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11014205.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11014205.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11014205.hg.1 | TC11001115.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1100170038.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100170038.hg.1 | ENST00000530578.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100170038.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100170038.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100170038.hg.1 | ENST00000530578.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100170038.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100170038.hg.1 | TC1100009330.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000525711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000525711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000525711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000526386.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000526386.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000526386.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000526386.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000526493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000526493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000526493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000526493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000529998.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000529998.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000529998.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000529998.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000529998.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000530578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000530578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000530578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000530578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000530917.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000530917.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000530917.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000530917.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000530917.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000530917.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000530917.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000533451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000533451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000533451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000533451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | ENST00000533451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009330.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100170037.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100170037.hg.1 | ENST00000530578.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100170037.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100170037.hg.1 | ENST00000284273.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100170037.hg.1 | ENST00000530578.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100170037.hg.1 | NM_032873.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100170037.hg.1 | TC1100009330.hg.1 | Clariom D TC |
