Assay Details
| Gene Symbol: LPAR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278000.1 | NP_001264929.1 | 4 - 5 | 542 | 61 | |
| XM_005262126.3 | XP_005262183.1 | 4 - 5 | 745 | 61 | ||
| XM_006724639.3 | XP_006724702.1 | 3 - 4 | 457 | 61 | ||
| XM_017029437.1 | XP_016884926.1 | 4 - 5 | 1804 | 61 | ||
| XM_017029438.1 | XP_016884927.1 | 2 - 3 | 480 | 61 | ||
| GenBank mRNA | AU129897.1 | - | 3 - 4 | 457 | 61 | |
| DA368656.1 | - | 4 - 5 | 525 | 61 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001278000.1,XM_005262126.3,XM_006724639.3,XM_017029437.1,XM_017029438.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000435339.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000435339.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000435339.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000435339.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000435339.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000435339.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000435339.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000514744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000514744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000514744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000514744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000514744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000607964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000607964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000607964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000607964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000610214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000610214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000610214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000610214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000610214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | NM_005296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | NM_005296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | NM_005296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | NM_005296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | NM_005296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | NM_005296.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007713.hg.1 | NM_005296.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00152853.hg.1 | ENST00000435339.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152853.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152853.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00152853.hg.1 | NM_005296.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00152853.hg.1 | ENST00000435339.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152853.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152853.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00152853.hg.1 | NM_005296.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00152853.hg.1 | TC0X00007713.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00152845.hg.1 | ENST00000514744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152845.hg.1 | ENST00000607964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152845.hg.1 | ENST00000610214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152845.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152845.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00152845.hg.1 | ENST00000514744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152845.hg.1 | ENST00000607964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152845.hg.1 | ENST00000610214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152845.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152845.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00152845.hg.1 | TC0X00007713.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00152852.hg.1 | ENST00000435339.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152852.hg.1 | ENST00000514744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152852.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152852.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00152852.hg.1 | NM_005296.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00152852.hg.1 | ENST00000435339.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152852.hg.1 | ENST00000514744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152852.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152852.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00152852.hg.1 | NM_005296.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00152852.hg.1 | TC0X00007713.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000435339.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000435339.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000435339.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000435339.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000435339.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000435339.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000435339.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000514744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000514744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000514744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000514744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000514744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000607964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000607964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000607964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000607964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000610214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000610214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000610214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000610214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000610214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | NM_005296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | NM_005296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | NM_005296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | NM_005296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | NM_005296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | NM_005296.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007713.hg.1 | NM_005296.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00152844.hg.1 | ENST00000607964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152844.hg.1 | ENST00000610214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152844.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152844.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00152844.hg.1 | ENST00000607964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152844.hg.1 | ENST00000610214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152844.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152844.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00152844.hg.1 | TC0X00007713.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00152851.hg.1 | ENST00000435339.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152851.hg.1 | ENST00000514744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152851.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152851.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00152851.hg.1 | NM_005296.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00152851.hg.1 | ENST00000435339.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152851.hg.1 | ENST00000514744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152851.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152851.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00152851.hg.1 | NM_005296.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00152851.hg.1 | TC0X00007713.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00152841.hg.1 | ENST00000435339.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152841.hg.1 | ENST00000514744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152841.hg.1 | ENST00000607964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152841.hg.1 | ENST00000610214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152841.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152841.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00152841.hg.1 | NM_005296.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00152841.hg.1 | ENST00000435339.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152841.hg.1 | ENST00000514744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152841.hg.1 | ENST00000607964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152841.hg.1 | ENST00000610214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152841.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152841.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00152841.hg.1 | NM_005296.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00152841.hg.1 | TC0X00007713.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00152843.hg.1 | ENST00000514744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152843.hg.1 | ENST00000607964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152843.hg.1 | ENST00000610214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152843.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152843.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00152843.hg.1 | ENST00000514744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152843.hg.1 | ENST00000607964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152843.hg.1 | ENST00000610214.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152843.hg.1 | ENST00000614823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00152843.hg.1 | NM_001278000.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00152843.hg.1 | TC0X00007713.hg.1 | Clariom D TC |

LPAR4
Genomic Map