Assay Details
| Gene Symbol: ZCCHC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024936.2 | NP_079212.2 | 11 - 12 | 1302 | 111 | |
| XM_011513835.1 | XP_011512137.1 | 12 - 13 | 1314 | 111 | ||
| XM_017008128.1 | XP_016863617.1 | 10 - 11 | 1181 | 111 | ||
| XM_017008129.1 | XP_016863618.1 | 9 - 10 | 1017 | 111 | ||
| GenBank mRNA | AK026677.1 | - | 6 - 7 | 571 | 111 | |
| AK299211.1 | - | 11 - 12 | 1292 | 111 | ||
| AY629351.1 | - | 11 - 12 | 1295 | 111 | ||
| BC016914.1 | - | 8 - 9 | 956 | 111 | ||
| BC157834.1 | - | 11 - 12 | 1296 | 9057053 | 111 | |
| BC171821.1 | - | 11 - 12 | 1296 | 9054475 | 111 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000175.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000580712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04002703.hg.1 | TC04000175.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04002703.hg.1 | ENST00000302874.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04002703.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04002704.hg.1 | TC04000175.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04002704.hg.1 | ENST00000302874.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04002704.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000580712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400148594.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148594.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148594.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148594.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400148594.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148594.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148594.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148594.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400148594.hg.1 | TC0400007095.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400148593.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148593.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148593.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148593.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148593.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400148593.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148593.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148593.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148593.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148593.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400148593.hg.1 | TC0400007095.hg.1 | Clariom D TC |

ZCCHC4
Genomic Map