Assay Details
| Gene Symbol: ZCCHC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318148.1 | NP_001305077.1 | 2 - 3 | 287 | 92 | |
| NM_024936.2 | NP_079212.2 | 2 - 3 | 287 | 92 | ||
| XM_011513835.1 | XP_011512137.1 | 2 - 3 | 254 | 92 | ||
| XM_017008129.1 | XP_016863618.1 | 2 - 3 | 254 | 92 | ||
| GenBank mRNA | AF161537.1 | - | 2 - 3 | 244 | 92 | |
| AK299211.1 | - | 2 - 3 | 277 | 92 | ||
| AK309675.1 | - | 2 - 3 | 276 | 92 | ||
| AY629351.1 | - | 2 - 3 | 280 | 92 | ||
| BC016914.1 | - | 2 - 3 | 266 | 92 | ||
| BC157834.1 | - | 2 - 3 | 281 | 9057053 | 92 | |
| BC171821.1 | - | 2 - 3 | 281 | 9054475 | 92 | |
| BG719159.1 | - | 2 - 3 | 292 | 92 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000175.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04002680.hg.1 | TC04000175.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04002680.hg.1 | ENST00000302874.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04002680.hg.1 | ENST00000450254.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04002680.hg.1 | ENST00000505451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04002680.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000580712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400148568.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148568.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148568.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148568.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148568.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400148568.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148568.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148568.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148568.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148568.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400148568.hg.1 | TC0400007095.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000302874.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000505451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000507760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000508058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000580712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | ENST00000612982.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007095.hg.1 | NM_024936.2 | Refseq |

ZCCHC4
Genomic Map