Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008242.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008242.hg.1 |
ENST00000370528.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008242.hg.1 |
ENST00000446890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008242.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008242.hg.1 |
TC10000708.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
ENST00000488090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
ENST00000488090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
ENST00000488090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008245.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008245.hg.1 |
ENST00000370528.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008245.hg.1 |
ENST00000370534.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008245.hg.1 |
ENST00000446890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008245.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008245.hg.1 |
TC10000708.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008247.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008247.hg.1 |
ENST00000370528.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008247.hg.1 |
ENST00000370534.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008247.hg.1 |
ENST00000446890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008247.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008247.hg.1 |
TC10000708.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008239.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008239.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008239.hg.1 |
TC10000708.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008241.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008241.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008241.hg.1 |
TC10000708.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008246.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008246.hg.1 |
ENST00000370528.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008246.hg.1 |
ENST00000370534.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008246.hg.1 |
ENST00000446890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008246.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008246.hg.1 |
TC10000708.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008243.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008243.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008243.hg.1 |
TC10000708.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008240.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008240.hg.1 |
ENST00000370528.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008240.hg.1 |
ENST00000446890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008240.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008240.hg.1 |
TC10000708.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008248.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008248.hg.1 |
ENST00000370528.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008248.hg.1 |
ENST00000370534.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008248.hg.1 |
ENST00000446890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008248.hg.1 |
ENST00000545665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008248.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008248.hg.1 |
TC10000708.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000158716.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158716.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158716.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158716.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158716.hg.1 |
TC1000008613.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
ENST00000488090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
ENST00000488090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
ENST00000488090.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008613.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158717.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158717.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158717.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158717.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158717.hg.1 |
TC1000008613.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000158715.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158715.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158715.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158715.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158715.hg.1 |
TC1000008613.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000158718.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158718.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158718.hg.1 |
ENST00000356713.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158718.hg.1 |
NM_020348.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158718.hg.1 |
TC1000008613.hg.1 |
Clariom D TC |