Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007058.hg.1 |
TC0X000528.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007058.hg.1 |
ENST00000262843.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007058.hg.1 |
ENST00000443968.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007058.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007058.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000443968.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000443968.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000443968.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000443968.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000443968.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000443968.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000443968.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000443968.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000443968.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000443968.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000443968.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000451923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000451923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000451923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000474517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000474517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000474517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
ENST00000474517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155228.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155228.hg.1 |
ENST00000443968.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155228.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155228.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155228.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155228.hg.1 |
ENST00000443968.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155228.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155228.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155228.hg.1 |
TC0X00008078.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000262843.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000443968.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000443968.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000443968.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000443968.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000443968.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000443968.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000443968.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000443968.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000443968.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000443968.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000443968.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000451923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000451923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000451923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000474517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000474517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000474517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
ENST00000474517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_012216.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008078.hg.1 |
NM_052817.2 |
Refseq |