Assay Details
Gene Symbol: MID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012216.3 | NP_036348.2 | 5 - 6 | 1644 | 150 | |
NM_052817.2 | NP_438112.2 | 5 - 6 | 1644 | 150 | ||
XM_005262062.4 | XP_005262119.1 | 5 - 6 | 5907 | 150 | ||
XM_017029239.1 | XP_016884728.1 | 5 - 6 | 5907 | 150 | ||
GenBank mRNA | AF196481.1 | - | 5 - 6 | 1084 | 150 | |
AY625004.1 | - | 4 - 5 | 1011 | 150 | ||
BC017707.1 | - | 4 - 5 | 1038 | 4446324 | 150 | |
BT006663.1 | - | 4 - 5 | 1011 | 150 | ||
Y18880.1 | - | 5 - 6 | 1268 | 150 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000528.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of autophagy
protein ubiquitination
negative regulation of viral transcription
protein localization to microtubule
positive regulation of I-kappaB kinase/NF-kappaB signaling
innate immune response
negative regulation of viral entry into host cell
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of NF-kappaB transcription factor activity
negative regulation of viral release from host cell
microtubule binding
zinc ion binding
ligase activity
protein homodimerization activity
protein heterodimerization activity
phosphoprotein binding
positive regulation of autophagy
protein ubiquitination
negative regulation of viral transcription
protein localization to microtubule
positive regulation of I-kappaB kinase/NF-kappaB signaling
innate immune response
negative regulation of viral entry into host cell
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of NF-kappaB transcription factor activity
negative regulation of viral release from host cell
microtubule binding
zinc ion binding
ligase activity
protein homodimerization activity
protein heterodimerization activity
phosphoprotein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR0X007062.hg.1 | TC0X000528.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007062.hg.1 | ENST00000262843.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007062.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007062.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007062.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007060.hg.1 | TC0X000528.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007060.hg.1 | ENST00000262843.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007060.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007060.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X007060.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000451923.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000451923.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000451923.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000474517.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000474517.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000474517.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | ENST00000474517.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155230.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155230.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155230.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155230.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155230.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155230.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155230.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155230.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155230.hg.1 | TC0X00008078.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000451923.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000451923.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000451923.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000474517.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000474517.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000474517.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | ENST00000474517.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008078.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155232.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155232.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155232.hg.1 | ENST00000474517.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155232.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155232.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155232.hg.1 | ENST00000262843.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155232.hg.1 | ENST00000443968.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155232.hg.1 | ENST00000474517.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00155232.hg.1 | NM_012216.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155232.hg.1 | NM_052817.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00155232.hg.1 | TC0X00008078.hg.1 | Clariom D TC |
