| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006836.hg.1 |
ENST00000230568.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006836.hg.1 |
ENST00000230568.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006836.hg.1 |
ENST00000230568.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006836.hg.1 |
ENST00000230568.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006836.hg.1 |
ENST00000230568.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006836.hg.1 |
ENST00000230568.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006836.hg.1 |
ENST00000379953.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006836.hg.1 |
ENST00000379953.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006836.hg.1 |
ENST00000379953.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006836.hg.1 |
ENST00000379953.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006836.hg.1 |
ENST00000379953.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006836.hg.1 |
ENST00000379953.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006836.hg.1 |
ENST00000379953.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006836.hg.1 |
NM_004271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006836.hg.1 |
NM_004271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006836.hg.1 |
NM_004271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006836.hg.1 |
NM_004271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006836.hg.1 |
NM_004271.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006836.hg.1 |
NM_004271.3 |
Refseq |