Assay Details
Gene Symbol: ARHGEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004706.3 | NP_004697.2 | 5 - 6 | 436 | 80 | |
NM_199002.1 | NP_945353.1 | 5 - 6 | 461 | 80 | ||
XM_005259386.4 | XP_005259443.1 | 5 - 6 | 788 | 80 | ||
XM_005259390.4 | XP_005259447.1 | 5 - 6 | 784 | 80 | ||
XM_011527468.2 | XP_011525770.1 | 5 - 6 | 784 | 80 | ||
GenBank mRNA | BC005155.2 | - | 2 - 3 | 174 | 80 | |
BC034013.1 | - | 5 - 6 | 392 | 3451036 | 80 | |
BQ944132.1 | - | 5 - 6 | 549 | 80 | ||
BT007421.1 | - | 2 - 3 | 150 | 80 | ||
U64105.1 | - | 5 - 6 | 381 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
