Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02003123.hg.1 |
TC02000213.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02003123.hg.1 |
ENST00000421745.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02003123.hg.1 |
NM_016252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200149579.hg.1 |
ENST00000421745.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200149579.hg.1 |
NM_016252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200149579.hg.1 |
ENST00000421745.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200149579.hg.1 |
NM_016252.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200149579.hg.1 |
TC0200007184.hg.1 |
Clariom D TC |