Assay Details
Gene Symbol: PDIK1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152835.4 | NP_690048.1 | 1 - 2 | 138 | 55 | |
GenBank mRNA | AK312872.1 | - | 1 - 2 | 138 | 55 | |
DB227864.1 | - | 1 - 2 | 138 | 55 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000333.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK312872.1,NM_152835.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | ENST00000374269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | ENST00000374269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | ENST00000374269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | ENST00000374269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | ENST00000374269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | ENST00000374271.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | ENST00000374271.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | ENST00000374271.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | ENST00000374271.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | ENST00000374271.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | ENST00000374271.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | ENST00000374271.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | ENST00000444713.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | ENST00000444713.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | ENST00000444713.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | ENST00000619836.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | ENST00000619836.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | ENST00000619836.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | ENST00000619836.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | ENST00000619836.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | ENST00000619836.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | NM_001243532.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | NM_001243532.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | NM_001243532.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | NM_001243532.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | NM_001243532.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | NM_001243532.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | NM_001243533.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | NM_001243533.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | NM_001243533.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | NM_001243533.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | NM_001243533.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | NM_001243533.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | NM_001243533.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | NM_001243533.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | NM_152835.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | NM_152835.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | NM_152835.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | NM_152835.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | NM_152835.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | NM_152835.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007439.hg.1 | NM_152835.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01005088.hg.1 | TC01000333.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01005088.hg.1 | ENST00000374269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01005088.hg.1 | ENST00000374271.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01005088.hg.1 | NM_001243533.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01005088.hg.1 | NM_152835.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01005085.hg.1 | TC01000333.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01005085.hg.1 | NM_001243533.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01005085.hg.1 | NM_152835.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151848.hg.1 | NM_001243533.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151848.hg.1 | NM_152835.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151848.hg.1 | NM_001243533.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151848.hg.1 | NM_152835.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151848.hg.1 | TC0100007439.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | ENST00000374269.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | ENST00000374269.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | ENST00000374269.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | ENST00000374269.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | ENST00000374269.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | ENST00000374271.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | ENST00000374271.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | ENST00000374271.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | ENST00000374271.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | ENST00000374271.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | ENST00000374271.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | ENST00000374271.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | ENST00000444713.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | ENST00000444713.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | ENST00000444713.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | ENST00000619836.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | ENST00000619836.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | ENST00000619836.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | ENST00000619836.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | ENST00000619836.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | ENST00000619836.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | NM_001243532.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | NM_001243532.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | NM_001243532.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | NM_001243532.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | NM_001243532.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | NM_001243532.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | NM_001243533.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | NM_001243533.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | NM_001243533.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | NM_001243533.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | NM_001243533.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | NM_001243533.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | NM_001243533.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | NM_001243533.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | NM_152835.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | NM_152835.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | NM_152835.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | NM_152835.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | NM_152835.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | NM_152835.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007439.hg.1 | NM_152835.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151851.hg.1 | ENST00000374269.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151851.hg.1 | ENST00000374271.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151851.hg.1 | NM_001243533.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151851.hg.1 | NM_152835.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151851.hg.1 | ENST00000374269.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151851.hg.1 | ENST00000374271.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100151851.hg.1 | NM_001243533.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151851.hg.1 | NM_152835.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100151851.hg.1 | TC0100007439.hg.1 | Clariom D TC |
