Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007216.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007216.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007216.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007216.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007216.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007216.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007216.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007216.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007216.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007216.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007216.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007216.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007216.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007216.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007216.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007216.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007216.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007216.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007216.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007216.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007216.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007216.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007216.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007216.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003948.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003948.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003948.hg.1 |
TC11000310.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003949.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003949.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003949.hg.1 |
TC11000310.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100150771.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150771.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150771.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150771.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150771.hg.1 |
TC1100007216.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007216.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007216.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007216.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007216.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007216.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007216.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007216.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007216.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007216.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007216.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007216.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007216.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007216.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007216.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007216.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007216.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007216.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007216.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007216.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007216.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007216.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007216.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007216.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007216.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150769.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150769.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150769.hg.1 |
ENST00000257836.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150769.hg.1 |
NM_024081.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150769.hg.1 |
TC1100007216.hg.1 |
Clariom D TC |