Assay Details
| Gene Symbol: HSPA14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016299.3 | NP_057383.2 | 8 - 9 | 972 | 87 | |
| GenBank mRNA | AF112210.1 | - | 8 - 9 | 791 | 87 | |
| AF143723.1 | - | 8 - 9 | 781 | 87 | ||
| AK225265.1 | - | 8 - 9 | 816 | 87 | ||
| AK226065.1 | - | 1 - 2 | 3645 | 87 | ||
| AK292323.1 | - | 8 - 9 | 780 | 87 | ||
| BC119690.1 | - | 8 - 9 | 791 | 40116697 | 87 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000112.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000441647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000441647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000441647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000441647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000441647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000470430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000470430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000470430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000470430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10001228.hg.1 | ENST00000378372.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10001228.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10001228.hg.1 | TC10000112.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10001229.hg.1 | ENST00000378372.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10001229.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10001229.hg.1 | TC10000112.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10001230.hg.1 | ENST00000378372.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10001230.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10001230.hg.1 | TC10000112.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000146942.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000146942.hg.1 | ENST00000470430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000146942.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000146942.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000146942.hg.1 | ENST00000470430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000146942.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000146942.hg.1 | TC1000006858.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000441647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000441647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000441647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000441647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000441647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000470430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000470430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000470430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000470430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NR_103464.1 | Refseq |

HSPA14
Genomic Map