Assay Details
| Gene Symbol: AIRE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000383.3 | NP_000374.1 | 9 - 10 | 1269 | 131 | |
| GenBank mRNA | AB006682.1 | - | 9 - 10 | 1227 | 131 | |
| AB006683.1 | - | 3 - 4 | 745 | 131 | ||
| AK309572.1 | - | 9 - 10 | 1831 | 131 | ||
| BC137268.1 | - | 2 - 3 | 926 | 9021265 | 131 | |
| BC137270.1 | - | 2 - 3 | 925 | 9021267 | 131 | |
| Z97990.1 | - | 9 - 10 | 1220 | 131 | ||
More Information
Set Membership: |
|

AIRE
Genomic Map