Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016254.hg.1 |
TC04001177.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016254.hg.1 |
ENST00000264319.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016254.hg.1 |
ENST00000358350.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016254.hg.1 |
ENST00000503238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016254.hg.1 |
ENST00000507711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016254.hg.1 |
ENST00000537810.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016254.hg.1 |
NM_015030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172551.hg.1 |
ENST00000358350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172551.hg.1 |
ENST00000503238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172551.hg.1 |
ENST00000507711.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172551.hg.1 |
NM_015030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172551.hg.1 |
ENST00000358350.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172551.hg.1 |
ENST00000503238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172551.hg.1 |
ENST00000507711.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172551.hg.1 |
NM_015030.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172551.hg.1 |
TC0400010626.hg.1 |
Clariom D TC |