Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008666.hg.1 |
ENST00000524610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008666.hg.1 |
ENST00000524610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008666.hg.1 |
ENST00000524610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008666.hg.1 |
ENST00000524610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008666.hg.1 |
ENST00000531034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008666.hg.1 |
ENST00000531034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008307.hg.1 |
TC06000697.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008307.hg.1 |
ENST00000370651.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008307.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008310.hg.1 |
TC06000697.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008310.hg.1 |
ENST00000370650.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008310.hg.1 |
ENST00000370651.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008310.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000473334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000473334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000473334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000473334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000578299.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000578299.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000578299.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000627002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000627002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000627002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000627002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000627002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
ENST00000627002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008308.hg.1 |
TC06000697.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008308.hg.1 |
ENST00000370644.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008308.hg.1 |
ENST00000370650.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008308.hg.1 |
ENST00000370651.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008308.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198973.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198973.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198973.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198973.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198973.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198973.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198973.hg.1 |
TC0600014142.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100008666.hg.1 |
ENST00000524610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008666.hg.1 |
ENST00000524610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008666.hg.1 |
ENST00000524610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008666.hg.1 |
ENST00000524610.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008666.hg.1 |
ENST00000531034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008666.hg.1 |
ENST00000531034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198968.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198968.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198968.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198968.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198968.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198968.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198968.hg.1 |
TC0600014142.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600198971.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198971.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198971.hg.1 |
ENST00000627002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198971.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198971.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198971.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198971.hg.1 |
ENST00000627002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198971.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198971.hg.1 |
TC0600014142.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600198970.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198970.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198970.hg.1 |
ENST00000627002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198970.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198970.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198970.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198970.hg.1 |
ENST00000627002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600198970.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600198970.hg.1 |
TC0600014142.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000370651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000473334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000473334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000473334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000473334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000578299.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000578299.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000578299.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000626021.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000627002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000627002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000627002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000627002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000627002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
ENST00000627002.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014142.hg.1 |
NM_003463.4 |
Refseq |