| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000371753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000371753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000371753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000371753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000371753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000371753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000371753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000371753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000371753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000371753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000371753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000371753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000467669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
ENST00000467669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011962.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900181789.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900181789.hg.1 |
ENST00000371753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900181789.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900181789.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900181789.hg.1 |
ENST00000371753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900181789.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900181789.hg.1 |
TC0900011962.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000277554.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000371753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000371753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000371753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000371753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000371753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000371753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000371753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000371753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000371753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000371753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000371753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000371753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000467669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
ENST00000467669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
NM_144653.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011962.hg.1 |
NM_144653.4 |
Refseq |