Assay Details
Gene Symbol: CMTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015050.2 | NP_055865.1 | 17 - 18 | 2074 | 84 | |
XM_005248955.3 | XP_005249012.1 | 17 - 18 | 2004 | 84 | ||
GenBank mRNA | AK292564.1 | - | 17 - 18 | 1928 | 84 | |
BC010731.2 | - | 2 - 3 | 201 | 84 | ||
BC031890.1 | - | 17 - 18 | 1968 | 4944457 | 84 | |
D43949.2 | - | 17 - 18 | 1964 | 84 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06000545.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06006019.hg.1 | TC06000545.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06006019.hg.1 | ENST00000373420.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06006019.hg.1 | ENST00000373451.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06006019.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000471097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000471097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000471097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000471097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000493656.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000493656.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000493656.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | ENST00000493656.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06006018.hg.1 | TC06000545.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06006018.hg.1 | ENST00000373420.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06006018.hg.1 | ENST00000373451.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06006018.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000455891.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000457419.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000471097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000471097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000471097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000471097.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000475364.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000493656.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000493656.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000493656.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | ENST00000493656.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007869.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600154718.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600154718.hg.1 | ENST00000475364.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600154718.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600154718.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600154718.hg.1 | ENST00000475364.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600154718.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600154718.hg.1 | TC0600007869.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600154719.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600154719.hg.1 | ENST00000475364.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600154719.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600154719.hg.1 | ENST00000373451.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600154719.hg.1 | ENST00000475364.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600154719.hg.1 | NM_015050.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600154719.hg.1 | TC0600007869.hg.1 | Clariom D TC |
