Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000229824.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007848.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154460.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154460.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154460.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154460.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154460.hg.1 |
TC0600007848.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600154462.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154462.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154462.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154462.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154462.hg.1 |
TC0600007848.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600154463.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154463.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154463.hg.1 |
ENST00000612677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154463.hg.1 |
NM_001257357.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154463.hg.1 |
TC0600007848.hg.1 |
Clariom D TC |