Assay Details
Gene Symbol: RAB44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257357.1 | NP_001244286.1 | 9 - 10 | 2676 | 64 | |
XM_011514600.2 | XP_011512902.1 | 9 - 10 | 2773 | 64 | ||
XM_011514601.2 | XP_011512903.1 | 10 - 11 | 2820 | 64 | ||
XM_011514602.2 | XP_011512904.1 | 9 - 10 | 2740 | 64 | ||
XM_011514604.2 | XP_011512906.1 | 9 - 10 | 2773 | 64 | ||
XM_017010855.1 | XP_016866344.1 | 9 - 10 | 4212 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600154472.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600154472.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600154472.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600154472.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600154472.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600154472.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600154472.hg.1 | TC0600007848.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600154473.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600154473.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600154473.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600154473.hg.1 | ENST00000229824.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600154473.hg.1 | ENST00000612677.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600154473.hg.1 | NM_001257357.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600154473.hg.1 | TC0600007848.hg.1 | Clariom D TC |
