Assay Details
| Gene Symbol: MRPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020236.3 | NP_064621.3 | 3 - 4 | 600 | 147 | |
| XM_011532198.2 | XP_011530500.1 | 4 - 5 | 652 | 147 | ||
| XM_011532199.1 | XP_011530501.1 | 4 - 5 | 466 | 147 | ||
| GenBank mRNA | AB049474.1 | - | 2 - 3 | 374 | 147 | |
| AF212225.1 | - | 3 - 4 | 502 | 149 | ||
| AK024791.1 | - | 1 - 2 | 203 | 147 | ||
| AK315600.1 | - | 3 - 4 | 417 | 147 | ||
| BC014356.1 | - | 3 - 4 | 424 | 3686618 | 147 | |
| BC015109.1 | - | 3 - 4 | 413 | 4044993 | 147 | |
| BC017765.1 | - | 3 - 4 | 421 | 147 | ||
| BC032595.1 | - | 3 - 4 | 411 | 5527878 | 147 | |
| CR457191.1 | - | 2 - 3 | 336 | 147 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000433.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04006138.hg.1 | TC04000433.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006138.hg.1 | ENST00000315567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006138.hg.1 | ENST00000538314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006138.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006139.hg.1 | TC04000433.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006139.hg.1 | ENST00000315567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006139.hg.1 | ENST00000538314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006139.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006140.hg.1 | TC04000433.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006140.hg.1 | ENST00000315567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006140.hg.1 | ENST00000538314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006140.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006145.hg.1 | TC04000433.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006145.hg.1 | ENST00000315567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006145.hg.1 | ENST00000506674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006145.hg.1 | ENST00000538314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006145.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006129.hg.1 | TC04000433.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006129.hg.1 | ENST00000315567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006129.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006136.hg.1 | TC04000433.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006136.hg.1 | ENST00000315567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006136.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006137.hg.1 | TC04000433.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006137.hg.1 | ENST00000315567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006137.hg.1 | ENST00000538314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006137.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006143.hg.1 | TC04000433.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006143.hg.1 | ENST00000315567.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006143.hg.1 | ENST00000506674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006143.hg.1 | ENST00000538314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04006143.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000502384.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000502384.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000502384.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000502384.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000502384.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000502384.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000502384.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000502384.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000504901.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000504901.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000504901.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000504901.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000504901.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000504901.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000504901.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000506674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000506674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000506674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000506674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000506674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000506674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000511521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000511521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000511521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000511521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | ENST00000515625.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007923.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400154404.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154404.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400154404.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154404.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400154404.hg.1 | TC0400007923.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400154414.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154414.hg.1 | ENST00000502384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154414.hg.1 | ENST00000506674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154414.hg.1 | ENST00000511521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154414.hg.1 | ENST00000515625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154414.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400154414.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154414.hg.1 | ENST00000502384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154414.hg.1 | ENST00000506674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154414.hg.1 | ENST00000511521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154414.hg.1 | ENST00000515625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154414.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400154414.hg.1 | TC0400007923.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400154416.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154416.hg.1 | ENST00000502384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154416.hg.1 | ENST00000506674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154416.hg.1 | ENST00000511521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154416.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400154416.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154416.hg.1 | ENST00000502384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154416.hg.1 | ENST00000506674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154416.hg.1 | ENST00000511521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154416.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400154416.hg.1 | TC0400007923.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000502384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000502384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000502384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000502384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000502384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000502384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000502384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000502384.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000504901.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000504901.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000504901.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000504901.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000504901.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000504901.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000504901.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000506674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000506674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000506674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000506674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000506674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000506674.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000511521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000511521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000511521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000511521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | ENST00000515625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007923.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400154411.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154411.hg.1 | ENST00000511521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154411.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400154411.hg.1 | ENST00000315567.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154411.hg.1 | ENST00000511521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400154411.hg.1 | NM_020236.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400154411.hg.1 | TC0400007923.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 128505 | - | - |

MRPL1
Genomic Map