Assay Details
| Gene Symbol: PIGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002641.3 | NP_002632.1 | 4 - 5 | 1093 | 70 | |
| NM_020473.3 | NP_065206.3 | 3 - 4 | 316 | 70 | ||
| NR_033835.1 | 4 - 5 | 719 | 70 | |||
| NR_033836.1 | 4 - 5 | 435 | 70 | |||
| XM_011545539.2 | XP_011543841.1 | 3 - 4 | 1917 | 70 | ||
| XM_017029581.1 | XP_016885070.1 | 3 - 4 | 1040 | 70 | ||
| GenBank mRNA | AK098047.1 | - | 3 - 4 | 529 | 70 | |
| AK303538.1 | - | 3 - 4 | 308 | 70 | ||
| AK315063.1 | - | 3 - 4 | 1052 | 70 | ||
| BC038236.1 | - | 3 - 4 | 1050 | 5733899 | 70 | |
| CX870731.1 | - | 3 - 4 | 395 | 70 | ||
| D11466.1 | - | 4 - 5 | 1062 | 70 | ||
| DA951065.1 | - | 4 - 5 | 435 | 70 | ||
| S74936.1 | - | 2 - 3 | 535 | 70 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000875.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000463173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000463173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000463173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000463173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000463173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000474662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000474662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000474662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000474662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000474662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000474662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000475746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000475746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000482148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000482148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000482148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000482148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000482148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000482148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000482148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000482148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000482148.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000542278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000542278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000542278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000542278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000542278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000542278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000542278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000542278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | ENST00000542278.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NM_020473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NM_020473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NM_020473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NM_020473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NM_020473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NM_020473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NM_020473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NM_020473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NM_020473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NR_033836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NR_033836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NR_033836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NR_033836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NR_033836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NR_033836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NR_033836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NR_033836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NR_033836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NR_033836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009124.hg.1 | NR_033836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011689.hg.1 | TC0X000875.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011689.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011689.hg.1 | ENST00000428964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011689.hg.1 | ENST00000482148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011689.hg.1 | ENST00000542278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011689.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011689.hg.1 | NM_020473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011689.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011689.hg.1 | NR_033836.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00162576.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00162576.hg.1 | ENST00000463173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00162576.hg.1 | ENST00000482148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00162576.hg.1 | ENST00000542278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00162576.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00162576.hg.1 | NM_020473.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00162576.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00162576.hg.1 | NR_033836.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00162576.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00162576.hg.1 | ENST00000463173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00162576.hg.1 | ENST00000482148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00162576.hg.1 | ENST00000542278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00162576.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00162576.hg.1 | NM_020473.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00162576.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00162576.hg.1 | NR_033836.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00162576.hg.1 | TC0X00009124.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000333590.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000463173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000463173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000463173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000463173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000463173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000474662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000474662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000474662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000474662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000474662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000474662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000475746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000475746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000482148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000482148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000482148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000482148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000482148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000482148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000482148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000482148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000482148.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000542278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000542278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000542278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000542278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000542278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000542278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000542278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000542278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | ENST00000542278.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NM_002641.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NM_020473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NM_020473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NM_020473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NM_020473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NM_020473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NM_020473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NM_020473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NM_020473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NM_020473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NR_033835.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NR_033836.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NR_033836.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NR_033836.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NR_033836.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NR_033836.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NR_033836.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NR_033836.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NR_033836.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NR_033836.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NR_033836.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009124.hg.1 | NR_033836.1 | Refseq |

PIGA
Genomic Map