Assay Details
Gene Symbol: KCNK15-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_132377.1 | 1 - 2 | 260 | 58 | ||
GenBank mRNA | BF195580.1 | - | 1 - 2 | 54 | 58 | |
DB261503.1 | - | 1 - 2 | 259 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000419931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000419931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000419931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000419931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000419931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000164355.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164355.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000164355.hg.1 | TC2000009199.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000164355.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164355.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000419931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000419931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000419931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000419931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000419931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000164362.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164362.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000164362.hg.1 | TC2000009199.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000164362.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164362.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000164357.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164357.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164357.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164357.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000164357.hg.1 | TC2000009199.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000164357.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164357.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164357.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164357.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000164351.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164351.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164351.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164351.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000164351.hg.1 | TC2000009199.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000164351.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164351.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164351.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164351.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000164356.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164356.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164356.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164356.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164356.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164356.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000164356.hg.1 | TC2000009199.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000164356.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164356.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164356.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164356.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164356.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164356.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000164361.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164361.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164361.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164361.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000164361.hg.1 | TC2000009199.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000164361.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164361.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164361.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164361.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000164350.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164350.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000164350.hg.1 | TC2000009199.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000164350.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000164350.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000164363.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000164363.hg.1 | TC2000009199.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000164363.hg.1 | NR_132377.1 | Refseq |
