Assay Details
Gene Symbol: KCNK15-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DB261503.1 | - | 2 - 3 | 380 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000419931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000419931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000419931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000419931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000419931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000255183.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000415889.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000419931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000419931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000419931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000419931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000419931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427303.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000427598.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | ENST00000445420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_034104.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_132377.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009199.hg.1 | NR_132377.1 | Refseq |
