Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR2000149967.hg.1 |
ENST00000202028.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149967.hg.1 |
ENST00000338074.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149967.hg.1 |
ENST00000373941.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149967.hg.1 |
ENST00000373946.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149967.hg.1 |
ENST00000373950.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149967.hg.1 |
ENST00000441639.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149967.hg.1 |
ENST00000479336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149967.hg.1 |
NM_001258329.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149967.hg.1 |
NM_001258331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149967.hg.1 |
NM_012156.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149967.hg.1 |
NM_177996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149967.hg.1 |
TC2000007239.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000149967.hg.1 |
ENST00000202028.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149967.hg.1 |
ENST00000338074.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149967.hg.1 |
ENST00000373941.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149967.hg.1 |
ENST00000373946.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149967.hg.1 |
ENST00000373950.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149967.hg.1 |
ENST00000441639.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149967.hg.1 |
ENST00000479336.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149967.hg.1 |
NM_001258329.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149967.hg.1 |
NM_001258331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149967.hg.1 |
NM_012156.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149967.hg.1 |
NM_177996.2 |
Refseq |