Assay Details
| Gene Symbol: CENPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022909.3 | NP_075060.1 | 5 - 6 | 457 | 132 | |
| GenBank mRNA | AB035124.1 | - | 5 - 6 | 370 | 132 | |
| AK290746.1 | - | 5 - 6 | 440 | 132 | ||
| BC012024.1 | - | 5 - 6 | 433 | 4510607 | 132 | |
| BC015355.1 | - | 5 - 6 | 439 | 3451767 | 132 | |
| KU178758.1 | - | 5 - 6 | 370 | 132 | ||
| KU178759.1 | - | 5 - 6 | 329 | 132 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000299.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000510742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000510742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000510742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003923.hg.1 | TC05000299.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003923.hg.1 | ENST00000283006.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003923.hg.1 | ENST00000515001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003923.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500151916.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151916.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151916.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151916.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151916.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500151916.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151916.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151916.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151916.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151916.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500151916.hg.1 | TC0500007666.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000283006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000502689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000510742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000510742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000510742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000513575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000514753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | ENST00000515001.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007666.hg.1 | NM_022909.3 | Refseq |

CENPH
Genomic Map